Learn More
TARGETMOL CHEMICALS INC LX-7101 hydrochloride 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T329851MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. LX-7101 hydrochloride is a potent LIMK and ROCK2 inhibitor with antihypertensive activity, inhibits LIMK1/2, ROCK, PKA, and can be used to study glaucoma. Purity 98.35%
Catalog No. 50-287-7804