Learn More
Medchemexpress LLC LX2343 | 333745-53-2 | 99.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HY11138310MM/1ML
Encompass_Preferred
LX2343 is a BACE1 enzyme inhibitor that also acts as a non-ATP competitive PI3K inhibitor. It stimulates autophagy to promote Aβ clearance.
  • BACE1 enzyme inhibitor with an IC50 value of 11.43±0.36 μM.
  • Non-ATP competitive PI3K inhibitor with an IC50 of 15.99±3.23 μM.
  • Stimulates autophagy.
  • Promotes Aβ clearance.
Catalog No. 50-003-43123