Learn More
Medchemexpress LLC LXR-623 100mg | 875787-07-8 | 100MG
SDP
Supplier:  Medchemexpress LLC HY10629100MG
Encompass_Preferred
LXR-623 is a brain-penetrant partial LXR and full LXR agonist with IC50s of 24 nM and 179 nM respectively
Catalog No. 50-002-42249