Learn More
Medchemexpress LLC HY-10629 5mg Medchemexpress, LXR-623 CAS:875787-07-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY106295MG
Encompass_Preferred
Medchemexpress, HY-10629 5mg LXR-623 CAS:875787-07-8 LXR-623 is a brain-penetrant partial LXRα and full LXRβ agonist, with IC50s of 24 nM and 179 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8335