Learn More
Apexbio Technology LLC LXR-623 875787-07-8 100mg
SDP
Supplier:  Apexbio Technology LLC B1264100
Encompass_Preferred
LXR-623 (CAS 875787-07-8) is a small-molecule partial agonist targeting liver X receptors LXR- and LXR- It selectively activates LXR- and LXR- modulating cholesterol homeostasis by regulating gene expression involved in cholesterol transport LXR-623 exerts its biological activity primarily through induction of cholesterol transporters such as ABCA1 and ABCG1 enhancing cholesterol efflux and reverse cholesterol transport In animal models oral administration of LXR-623 increases expression of ABCA1 and ABCG1 in peripheral blood cells and intestinal ABCG5/ABCG8 in mouse atherosclerosis models with reported IC50 values of 24 nM for LXR- and 179 nM for LXR- Based on these pharmacological properties LXR-623 holds research potential in atherosclerosis and Alzheimer s disease
Catalog No. 50-000-92192