Learn More
TARGETMOL CHEMICALS INC LY 2389575 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T222045MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LY2389575 hydrochloride is an mGlu3 negative allosteric modulator (NAM) that is selective and non-competitive with an IC50 value of 190 nM. LY2389575 hydrochloride induces an increase in the level of Mrc1 which amplifies the toxicity of (beta)-amyloid (A(beta)) and can be used to study Alzheimer's disease. It can be used to study Alzheimer's disease. purity: 100%
Catalog No. 50-258-0970