Learn More
Medchemexpress LLC Ly-2584702 tosylate | 1082949-68-5 | 99.5% | 617.6 g/mol | C28H27F4N7O3S | 250MG
SDP
Supplier:  Medchemexpress LLC HY12493A250MG
Encompass_Preferred
LY-2584702 tosylate is the tosylate salt of LY-2584702, a selective, ATP-competitive inhibitor of p70 ribosomal S6 kinase (p70S6K/S6K1) used as a research tool in biochemical, cellular, and preclinical studies. Reported biochemical IC50 is 4 nM, S6K1 enzyme assay IC50 ≈ 2 nM, and cellular pS6 inhibition is observed at approximately 100 nM. High-purity material (CAS 1082949-68-5) is supplied for laboratory use.
  • Selective ATP-competitive inhibitor of p70S6K.
  • Biochemical potency: IC50 = 4 nM; S6K1 enzyme assay IC50 ≈ 2 nM.
  • Demonstrated cellular pS6 inhibition at ~100 nM.
  • Suitable for biochemical, cellular, and in vivo pharmacology studies.
  • High purity (99.5%) and characterized by molecular weight 617.6 g/mol.
  • Provided as the tosylate salt (CAS 1082949-68-5) for stability and handling.
Catalog No. 50-002-37250