Learn More
Medchemexpress LLC Ly-2584702 tosylate | 1082949-68-5 | MFCD29054739 | 99.5% | 617.62 g/mol | C28H27F4N7O3S | 25MG
SDP
Supplier:  Medchemexpress LLC HY12493A25MG
Encompass_Preferred
LY-2584702 tosylate salt is a selective, ATP-competitive small-molecule inhibitor of p70S6K (S6K1) used in biochemical, cell-based, and in vivo preclinical research. It exhibits low-nanomolar enzyme potency and reduces phosphorylation of the S6 ribosomal protein in cells; it has demonstrated efficacy in xenograft models.
Catalog No. 50-002-35197