Learn More
Medchemexpress LLC Ly-2584702 tosylate salt | 1082949-68-5 | 99.5% | 617.62 | C28H27F4N7O3S | 1G

Supplier: Medchemexpress LLC HY12493A1G
LY-2584702 tosylate salt is an ATP-competitive inhibitor of p70S6K (S6K1) with nanomolar enzyme potency and reported cellular activity; intended for research use only.
- Selective ATP-competitive p70S6K inhibitor (enzyme IC50 = 4 nM).
- S6K1 enzyme assay IC50 = 2 nM.
- Inhibits phosphorylation of ribosomal protein S6 in cells (pS6 IC50 ≈ 0.1-0.24 μM).
- High purity: 99.5%.
- Molecular weight 617.62 g/mol.
- Storage: 4 °C sealed; in solvent -80 °C (1 year), -20 °C (6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.