Learn More
Medchemexpress LLC Ly-2584702 tosylate salt | 1082949-68-5 | 99.5% | 617.62 | C28H27F4N7O3S | 500MG

Supplier: Medchemexpress LLC HY12493A500MG
LY-2584702 tosylate salt is a selective ATP-competitive inhibitor of p70S6K used for biochemical and cellular research into S6K signaling. It exhibits low-nanomolar potency in enzyme assays and reduces phosphorylation of S6 in cellular models. The compound is supplied as a light yellow solid and is typically prepared as a DMSO stock for in vitro studies.
- Selective ATP-competitive inhibitor of p70S6K; p70S6K IC50 4 nM, S6K1 enzyme assay IC50 2 nM.
- Inhibits cellular pS6 with effective concentrations around 100 nM; HCT116 pS6 IC50 0.1-0.24 μM.
- Observed off-target activity against MSK2 and RSK at higher concentrations (58-176 nM).
- High purity (99.5%) and light yellow solid appearance.
- Molecular formula C28H27F4N7O3S; molecular weight 617.62 g/mol.
- Stable when stored sealed away from moisture; in solution store at -80°C (1 year) or -20°C (6 months).
- Soluble in DMSO; prepare a 10 mM stock solution for in vitro assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.