Learn More
Medchemexpress LLC LY-2584702 tosylate salt | 1082949-68-5 | MFCD29054739 | 99.5% | 617.62 | C28H27F4N7O3S | 50MG
SDP
Supplier:  Medchemexpress LLC HY12493A50MG
Encompass_Preferred
LY-2584702 tosylate salt is a small-molecule, ATP-competitive inhibitor of p70 ribosomal S6 kinase (p70S6K/S6K1) supplied as the tosylate salt for research use only. It exhibits low-nanomolar enzyme potency and cellular inhibition of S6 phosphorylation, making it suitable for studies of S6K signaling and related pathways.
  • Selective ATP-competitive inhibition of p70S6K.
  • Low-nanomolar enzyme potency (IC50 ≈ 2-4 nM).
  • Cellular inhibition of S6 phosphorylation (IC50 ≈ 0.1-0.24 μM).
  • Provided as a tosylate salt for improved handling.
  • High reported purity (99.5%).
  • Intended for laboratory research use only.
Catalog No. 50-002-37251