Learn More
TARGETMOL CHEMICALS INC LY-2584702 TOSYLATE SALT 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T174610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY-2584702 tosylate salt is a selective ATP-competitive p70S6K inhibitor used in trials studying the treatment of cancer. purity: 98%
Catalog No. 50-257-7310