Learn More
TARGETMOL CHEMICALS INC LY-2624803 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T244215MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LY-2624803 is a 5-HT2A receptor antagonist and histamine receptor inverse agonist that can be used to improve slow wave sleep. purity: 99%
Catalog No. 50-258-0810