Learn More
AdipoGen LY-333,531 . hydrochloride (5 mg)
SDP
Supplier:  AdipoGen AGCR10087M005
Encompass_Preferred
Chemical. CAS: 169939-93-9. Formula: C28H28N4O3 . HCl. MW: 468.6 . 36.5. Isozyme selective inhibitor of protein kinase Cbeta (PKCbeta). PKCbetaI and PKCbetaII isozyme inhibitor. Amelioriates diabetic retinopathy, diabetic peripheral neuropathy and diabetic nephropathy. Anti-cancer and anti-angiogenic compound. Suppresses glucose-induced adhesion of human monocytes to endothelial cells. Suppresses ERK1/2 and Akt phosphorylation.
Catalog No. 50-205-3827