Learn More
Apexbio Technology LLC LY-411575, 10mg. Cas: 209984-57-6 MFCD: MFCD09832551. Gamma secretase inhibitor
SDP
Supplier:  Apexbio Technology LLC A401910
Encompass_Preferred
LY-411575 is a potent inhibitor of -secretase with IC50 value of 0.078 nM in MEM assay.[1]-Secertase is one of intramenbrane-cleaving aspartyl protease which cleaves many type-I MEM proteins and many of them have important biological functions. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4858