Learn More
Medchemexpress LLC LY-411575 (isomer 1) | 209984-58-7 | 99.5% | C26H23F2N3O4 | 5 MG

Supplier: Medchemexpress LLC HY50752A5MG
LY-411575 isomer 1 is an isomer of LY411575, which is a potent γ-secretase inhibitor. It has a molecular weight of 479.48 and appears as a solid, white to off-white in color.
- Potent γ-secretase inhibitor
- Appears as a solid
- White to off-white in color
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 2 years or -20°C for 1 year
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.