Learn More
Apexbio Technology LLC LY2228820, 100mg. Cas: 862507-23-1 MFCD: MFCD22572532. P38 MAP kinase inhibitor
SDP
Supplier:  Apexbio Technology LLC A5566100
Encompass_Preferred
LY2228820 is a potent, selective, ATP-competitive small-molecule inhibitor inhibiting - and -isoforms of p38 MAPK with IC(50) values of 5.3nM and 3.2 nM, respectively1. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4955