Learn More
Cayman Chemical LY2365109hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 3742710 MG
Encompass_Preferred
An inhibitor of GlyT1a (IC50 = 15.8 nM in BE2-C cells expressing the human transporter); selective for GlyT1a over GlyT2 (IC50 = >30 µM in RD cells expressing the human transporter); induces locomotor hyperactivity, impaired gait, compulsive walking, and labored breathing, as well as impairs the ability to stand, in rats at 10 mg/kg; prevents maximal electroshock-induced acute seizures in mice at 30 mg/kg and decreases seizure number in an electroshock-induced rat model of chronic epilepsy at 10 mg/kg
Catalog No. 50-291-7526