Learn More
Medchemexpress LLC Ly2389575 hydrochloride | 885104-09-6 | MFCD28133385 | 99.9% | 438.58 | C15H16BrCl3N4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10750925MG
Encompass_Preferred
LY2389575 hydrochloride is a research-only small molecule that acts as a selective, non-competitive negative allosteric modulator of the metabotropic glutamate receptor 3 (mGlu3). It is used to probe mGlu3 pharmacology and related pathways, including effects on Mrc1 levels and amyloid beta-related toxicity in preclinical studies.
  • Selective non-competitive mGlu3 negative allosteric modulator with reported IC50 ≈ 190 nM.
  • High reported purity suitable for biochemical and cellular assays.
  • Available in small milligram quantities and solution formats for assay flexibility.
  • Stable under recommended storage conditions for extended sample retention.
  • Useful for studies of receptor modulation, signaling, and amyloid beta interactions.
Catalog No. 50-002-41270