Learn More
Apexbio Technology LLC LY2409881 25mg. 524-95-8. MFCD00014823
SDP
Supplier:  Apexbio Technology LLC B498625
Encompass_Preferred
LY2409881 is a potent and selective inhibitor of IKK2 with IC50 value of 30 nM. IκB kinase β (IKK2) is an enzyme that serves as a subunit of IκB kinase, which is involved in the cytokine-activated immune responses. IKK2 activates nuclear factor kappa-B kinase (NF-κB). LY2409881 is a selective IKK2 inhibitor and exhibited >10-fold selectivity over other common kinases and IKK1. In the ovarian cancer cell line SKOV3, LY2409881 showed moderate cytotoxicity. While coadministration of LY2409881 and TNFɑ significantly induced cells death, which suggested LY2409881 inhibited NF-κB mediated antiapoptotic signals in a TNFɑ-dependent way. In the DLBCL cell line LY10, LY2409881 (10 μM) inhibited TNFɑ-dependent phosphorylation of IκB. In the DLBCL cell line SUDHL2, LY2409881 (10 μM) inhibited the nuclear signals of p50 and NF-κB. Also, LY2409881 induced apoptosis in a concentration-dependent way. Other sizes are available. Please inqury us for quote.
Catalog No. 50-115-2102