Learn More
Selleck Chemical LLC LY2510924 2mg 1088715-84-7
SDP
Supplier:  Selleck Chemical LLC S85052MG
Encompass_Preferred
LY2510924 is a potent and selective CXCR4 antagonist that specifically blocks SDF-1 binding to CXCR4 with IC50 value of 0.079 nmol/L and inhibits SDF-1-induced GTP binding with Kb value of 0.38 nmol/L. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8343