Learn More
Selleck Chemical LLC LY2584702 Tosylate
SDP
Supplier:  Selleck Chemical LLC S770450MG
Encompass_Preferred
LY2584702 Tosylate is a selective ATP-competitive p70S6K inhibitor with IC50 of 4 nM Phase 1
Catalog No. 50-000-00156