Learn More
TARGETMOL CHEMICALS INC LY266097 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T229475MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY266097 hydrochloride (LY 266097 hydrochloride) is an antagonist of 5-HT2 with pKis of 7.7 9.8 and 7.6 for human 5-HT2A 5-HT2B and 5-HT2C and can be used in studies about depression. purity: 99%
Catalog No. 50-258-0909