Learn More
Medchemexpress LLC Ly2794193 | 2173037-97-1 | >98.0% | 1 ML

Supplier: Medchemexpress LLC HY11924310MM 1ML
LY2794193 is a potent and selective metabotropic glutamate receptor 3 (mGlu3) agonist supplied as a ready-to-use stock solution for research applications.
- Potent and selective mGlu3 receptor agonist (hmGlu3 Ki = 0.927 nM; EC50 = 0.47 nM).
- Provided as a 10 mM solution, 1 mL, suitable for preparing working stocks.
- High reported purity: ≥98%.
- Molecular formula C16H18N2O6; molecular weight ~334.32 g·mol⁻¹.
- Soluble in DMSO; stock solutions commonly prepared in DMSO for in vitro assays.
- Intended for research use in pharmacology and receptor characterization studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.