Learn More
Apexbio Technology LLC LY2835219 1231930-82-7 100mg
SDP
Supplier:  Apexbio Technology LLC A1794100
Encompass_Preferred
LY2835219 (CAS 1231930-82-7) is a selective orally bioavailable inhibitor of cyclin-dependent kinases 4 and 6 (CDK4/6) It targets CDK4 and CDK6 with reported IC50 values of 2 nM and 10 nM respectively thereby reducing phosphorylation of the retinoblastoma protein (Rb) This mechanism leads to cell-cycle arrest during the G1 phase and subsequent inhibition of tumor cell proliferation LY2835219 demonstrates the capacity to cross the blood-brain barrier indicating its potential utility in research involving primary or metastatic intracranial tumors
Catalog No. 50-000-62783