Learn More
Selleck Chemical LLC LY3009120
SDP
Supplier:  Selleck Chemical LLC S78421G
Encompass_Preferred
LY03009120 (DP-4978) is a potent pan-Raf inhibitor with IC50 of 44 nM 31-47 nM and 42 nM for A-raf B-Raf and C-Raf in A375 cells respectively LY03009120 induces autophagy Phase 1
Catalog No. 50-000-00740