Learn More
Selleck Chemical LLC Crenigacestat (LY3039478) 5mg 1421438-81-4
SDP
Supplier:  Selleck Chemical LLC S71695MG
Encompass_Preferred
Crenigacestat (LY3039478) is an oral Notch and gamma-secretase inhibitor with IC50 of 0.41 nM for Notch. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4477