Learn More
Selleck Chemical LLC LY3214996 2mg 1951483-29-6
SDP
Supplier:  Selleck Chemical LLC S85342MG
Encompass_Preferred
LY3214996 is a selective and novel ERK1/2 inhibitor with IC50 of 5 nM for both enzymes in biochemical assays. It potently inhibits cellular phospho-RSK1 in BRAF and RAS mutant cancer cell lines. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8013