Learn More
Selleck Chemical LLC LY3295668 5mg
SDP
Supplier:  Selleck Chemical LLC S87825MG
Encompass_Preferred
LY3295668 (AK-01) is a potent, orally active and specific inhibitor of Aurora A kinase with Ki of 0.8 nM and 1038 nM for AURKA and AURKB, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3390
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.