Learn More
Medchemexpress LLC LY3509754 | 2452464-73-0 | 99.4% | 586.51 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY139206100MG
Encompass_Preferred
LY3509754, also known as IL-17A inhibitor 1, is a potent IL-17A inhibitor with demonstrated IC50 values of less than 9.45 nM in alphalisa assay and 9.3 nM in HT-29 cells. It exhibits high oral bioavailability (F=107%), making it a promising candidate for research.
  • Potent IL-17A inhibitor
  • High oral bioavailability (F=107%)
  • Effective in alphalisa assay and HT-29 cells
  • Supplied as 100 MG
Catalog No. 50-003-51300