Learn More
Medchemexpress LLC LY3522348 | 2568608-48-8 | 98.3% | 409.41 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14998750MG
Encompass_Preferred
LY3522348 (KHK-IN-3) is an inhibitor of ketohexokinase (KHK), a rate-limiting enzyme and fructokinase involved in fructose metabolism. KHK catalyzes the phosphorylation of fructose to fructose-1-phosphate (FIP) at the expense of ATP. The lack of feedback inhibition in fructose metabolism leads to the accumulation of downstream intermediates, contributing to lipogenesis, gluconeogenesis, and oxidative phosphorylation.
  • KHK inhibitor.
  • Can be utilized in studies related to kidney disease.
  • Applicable in research concerning nonalcoholic steatohepatitis (NASH).
  • Useful in diabetes research.
  • Can be used to study heart failure.
Catalog No. 50-003-60483