Learn More
Medchemexpress LLC Lyciumin A | 125708-06-7 | 98.59% | 873.91 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN95281MG
Encompass_Preferred
Lyciumin A is a cyclic octapeptide that inhibits proteases, renin, and angiotensin-converting enzyme, making it suitable for hypertension research. It can be utilized for research related to hypertension.
  • Inhibits proteases, renin, and angiotensin-converting enzyme
  • Useful for hypertension research
Catalog No. 50-004-26341