Learn More
Selleck Chemical LLC Lycopodii Herba Extract E3629-5MG
SDP
Supplier:  Selleck Chemical LLC E36295MG
Encompass_Preferred
Lycopodii Herba Extract is extracted from Lycopodii Herba the dried whole plant of Lycopodium japonicum Thunb (Lycopodiaceae) Lycopodii Herba possesses anti-inflammation activity antioxidative activity and antitumor activity and has been traditionally used for the prevention and treatment of various diseases such as rheumatoid arthritis contusion quadriplegia dysmenorrhea
Catalog No. 50-000-40402