Learn More
Medchemexpress LLC HY-N0288 50mg Medchemexpress, Lycorine CAS:476-28-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN028850MG
Encompass_Preferred
Medchemexpress, HY-N0288 50mg Lycorine CAS:476-28-8 Lycorine is a natural alkaloid extracted from the Amaryllidaceae plant family with antiviral, antimalarial and antiinflammation activities. Lycorine inhibits the growth and metastasis of hormone-refractory anti-prostate cancer (PCa) and induces cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0249