Learn More
Medchemexpress LLC Lymphocyte-specific protein 1 (human) | >95.0% | 50 UG
SDP
Supplier:  Medchemexpress LLC HYP7591650UG
Encompass_Preferred
Recombinant full-length human lymphocyte-specific protein 1 (LSP1) expressed in HEK293 cells and supplied with a C-terminal 6xHis tag. The protein binds actin and is implicated in cytoskeletal regulation, neutrophil activation, and chemotaxis. Supplied lyophilized at high purity for biochemical and cell biology research applications.
  • Expressed in HEK293 cells.
  • C-terminal 6xHis tag for detection and purification.
  • Full-length isoform corresponding to residues M1-P339.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation with stabilizers; store at -20 °C.
  • Suitable for biochemical and cell biology studies.
Catalog No. 50-002-91951