Learn More
Cosmo Bio Usa Inc Lymphoprep 250ml
SDP
Supplier:  Cosmo Bio Usa Inc AXS1114544
Encompass
Lymphoprep™ is a ready-made, sterile, and endotoxin tested solution that has been used to isolate mononuclear cells for more than 35 years. Mononuclear cells (monocytes and lymphocytes) have a lower buoyant density than erythrocytes and polymorphonuclear (PMN) leukocytes (granulocytes). The vast majority of mononuclear cells have densities below 1.077 g/ml. These cells can therefore be isolated by centrifugation on an iso-osmotic medium with a density of 1.077 g/ml. Mononuclear cells are therefore retained at the sample/medium interface while erythrocytes and PMNs sediment through the medium.This method is rapid, simple, and reliable and gives excellent results with blood samples from normal individuals and patients.
Catalog No. NC0343649