Learn More
Medchemexpress LLC Lyn-1604 dihydrochloride | 2310109-38-5 | 99.6% | 657.54 g/mol | C33H45Cl4N3O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY101923B10MM 1ML
Encompass_Preferred
LYN-1604 dihydrochloride is a research-grade small-molecule activator of UNC-51-like kinase 1 (ULK1) used to probe autophagy and cell death pathways in preclinical and in vitro studies, including triple-negative breast cancer models. The compound is supplied as a dihydrochloride salt in solution or solid forms for flexible experimental use.
  • Potent ULK1 activation with EC50 18.94 nM.
  • Available as 10 mM solution in DMSO and multiple solid sizes for convenient dosing.
  • High purity material (99.6%).
  • Accompanied by datasheet, certificate of analysis, and safety data sheet.
  • Useful for studies of autophagy, apoptosis, and cancer biology.
Catalog No. 50-002-48868