Learn More
Medchemexpress LLC Lyn peptide inhibitor | 222018-18-0 | 99.35% | 2370.88 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP11111MG
Encompass_Preferred
Lyn peptide inhibitor is a potent and cell-permeable inhibitor of the Lyn-coupled IL-5 receptor signaling pathway, while keeping other signals intact. It blocks Lyn activation and inhibits the binding of Lyn tyrosine kinase to the βc subunit of IL-3/GM-CSF/IL-5 receptors. It can be used for the study of asthma, allergic, and other eosinophilic disorders.
  • Potent and cell-permeable inhibitor
  • Blocks Lyn activation
  • Inhibits binding to βc subunit of IL-3/GM-CSF/IL-5 receptors
  • Useful for studying asthma, allergic, and eosinophilic disorders
Catalog No. 50-004-24664