Learn More
Selleck Chemical LLC Lynestrenol S4883-25mg
SDP
Supplier:  Selleck Chemical LLC S488325MG
Encompass_Preferred
Lynestrenol (Ethinylestrenol) is a synthetic progestational hormone an agonist of the progesterone receptor and has weak androgenic and estrogenic activity
Catalog No. 50-310-9196