Learn More
Meridian Life Science Lyo-Ready Direct DNA qPCR, 5ML
SDP
Supplier:  Meridian Life Science MDX1525
Encompass_Preferred
Lyo-Ready DNA qPCR Urine is a glycerol-free, high-concentration, ready-to-use qPCR formulation engineered with a proprietary buffer system designed for overcoming inhibitors found in urine. It allows accurate target amplification direct from urine or pooled urine samples. It can be used in multiplex assays that require minimal sample processing and fast turn-around times without compromising sensitivity, for low copy number samples. It also increase reproducibility in extracted samples, as extraction methods are not always 100% efficient, often resulting in loss of target nucleic acid and potential co-purification of urine PCR inhibitors that can interfere with amplification, leading to false-negative results. It is ideal for room temperature, lyophilized, singleplex and multiplex qPCR assays and is suited for high-throughput, automated platforms.
Catalog No. 50-008-56464