Learn More
MOLTOX Molecular Toxicology Inc 60-251.4L - Lyophilized NADPH REGENSYS B Plus. Intended for use in EAT testing. Supports 5-30% S9 mix. 40ml refers to final volume of S9 mix/pkg. 140mg of crystalline NADP. Add to A Plus. Store in freezer.
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 60251.4L
Encompass
60-251.4L - Lyophilized NADPH REGENSYS B Plus. Intended for use in EAT testing. Supports 5-30% S9 mix. 40ml refers to final volume of S9 mix/pkg. 140mg of crystalline NADP. Add to A Plus. Store in freezer.
Catalog No. NC3235405