Learn More
MOLTOX Molecular Toxicology Inc 60-251.5L - Lyophilized NADPH REGENSYS B Plus. Intended for use in EAT testing. Supports 5-30% S9 mix. 50ml refers to final volume of S9 mix/pkg. 175mg of crystalline NADP. Add to A Plus. Store in freezer.
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 60251.5L
Encompass
60-251.5L - Lyophilized NADPH REGENSYS B Plus. Intended for use in EAT testing. Supports 5-30% S9 mix. 50ml refers to final volume of S9 mix/pkg. 175mg of crystalline NADP. Add to A Plus. Store in freezer.
Catalog No. NC3235406