Learn More
Medchemexpress LLC Lys-Ala-pNA | 115846-24-7 | 99.8% | 337.37 | 10 MG

Supplier: Medchemexpress LLC HYP446810MG
Lys-Ala-pNA is a hydrolytic substrate for dipeptidyl peptidase (DPP) II, featuring a Km of 0.42 mM. It appears as a solid, ranging from light yellow to yellow in color. For research use only.
- Hydrolytic substrate of dipeptidyl peptidase (DPP) II
- Appearance: Solid
- Color: Light yellow to yellow
- Sequence: Lys-Ala-{pNA}
- Sequence shortening: KA-{pNA}
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.