Learn More
Medchemexpress LLC Lys-Arg-Thr-Leu-Arg-Arg acetate | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP2673A5MG
Encompass_Preferred
Lys-Arg-Thr-Leu-Arg-Arg acetate (KRTLRR acetate) is a hexapeptide. It is a substrate of EGF receptor protein kinase C and can be used to measure protein kinase C activity.
  • Hexapeptide
  • Substrate of EGF receptor protein kinase C
  • Used to measure protein kinase C activity
  • Soluble in H2O at ≥ 100 mg/mL
Catalog No. 50-004-00350