Learn More
Medchemexpress LLC Lys-Arg-Thr-Leu-Arg-Arg acetate | 829.01 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP2673A10MG
Encompass_Preferred
Lys-Arg-Thr-Leu-Arg-Arg (KRTLRR) acetate is a hexapeptide. It functions as a substrate of EGF receptor protein kinase C and can be utilized to measure protein kinase C activity.
  • Hexapeptide
  • Substrate of EGF receptor protein kinase C
  • Can be used to measure protein kinase C activity
  • For research use only
Catalog No. 50-004-00331