Learn More
Medchemexpress LLC Lys-bradykinin trifluoroacetate | 99.9% | 1188.4 g/mol | C56H85N17O12 · x C2HF3O2 | 5 MG

Supplier: Medchemexpress LLC HY103293A5MG
Lys-bradykinin trifluoroacetate is the trifluoroacetic acid (TFA) salt form of the peptide Lys-Bradykinin (kallidin 380-389). It is supplied as a purified powder for research applications where it serves as a ligand for kallidin and bradykinin receptors. Reported purity is 99.85% and the free-base molecular weight is 1188.38 g/mol.
- TFA salt of Lys-Bradykinin peptide.
- High purity (99.85%).
- Provided as a solid powder suitable for dissolution in common solvents.
- Recommended storage: sealed, protected from moisture and light; powder stable at -80°C for up to 2 years.
- Intended for research use as a ligand in receptor binding or signaling studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.