Learn More
Medchemexpress LLC HY-103295A 5mg Medchemexpress, Lys-[Des-Arg9]Bradykinin (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103295A5MG
Encompass_Preferred
Medchemexpress, HY-103295A 5mg Lys-[Des-Arg9]Bradykinin (TFA) CAS: Lys-[Des-Arg9]Bradykinin TFA, a naturally occurring kinin, is a potent and highly selective bradykinin B1 receptor agonist with a Ki of 0.12 nM, 1.7 nM and 0.23 nM for human, mouse and rabbit B1 receptors, respectively. Lys-[Des-Arg9]Bradykinin TFA has low inhibitory activity on B2 receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0665