Learn More
Medchemexpress LLC LYS006 | 1799681-85-8 | 98.4% | 392.77 | 1 ML

Supplier: Medchemexpress LLC HY13729810MM/1ML
LYS006 (LTA4H-IN-1) is a potent inhibitor of leukotriene A4 hydrolase (LTA4H) with an IC50 of 2 nM. It is utilized in research for inflammatory and autoimmune disorders.
- Potent LTA4H inhibitor (IC50 of 2 nM)
- Inhibits LTB4 biosynthesis in human whole blood assay (IC50 of 167 nM)
- Used for inflammatory and autoimmune disorder research
- Molecular weight: 392.77
- Appearance: White to off-white solid
- Storage: -20°C for powder, -80°C in solvent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.