Learn More
Medchemexpress LLC Lys006 hydrochloride | 2619563-79-8 | 429.23 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13729850MG
Encompass_Preferred
LYS006 hydrochloride is a potent inhibitor of leukotriene A4 hydrolase (LTA4H) with an IC50 of 2 nM. It is extracted from patent WO2015092740A1, example 29, and is for research use only. It can be used for the research of inflammatory and autoimmune disorders.
  • Potent inhibitor of LTA4H hydrolase
  • IC50 of 2 nM
  • Used for research on inflammatory and autoimmune disorders
  • Currently in Phase 2 clinical trials for inflammatory acne, ulcerative colitis, hidradenitis suppurativa, and non-alcoholic fatty liver disease/nonalcoholic steatohepatitis
Catalog No. 50-003-50678