Learn More
CPC Scientific H-Ala-Thr-Lys-Ala-Ala-Arg-Lys(Me1)-Ser-Ala-Pro-Ala-Thr-Gly-Gly-Val-Lys-Lys-Pro-His-Arg-Tyr-Arg-Pro-Gly-Gly-Lys(Biotin)-OH (trifluoroacetate salt) 0.25MG
SDP
Supplier:  CPC Scientific HIST053A
Encompass
This peptide is the fragment of Histone H3 (21-44) monomethylated at Lys27 with an additional C-terminal Gly followed by a biotinylated Lys.ONE-LETTER SEQUENCE: ATKAAR-K(Me1)-SAPATGGVKKPHRYRPG-GK(Biotin)MOLECULAR FORMULA: MOLECULAR WEIGHT:2931.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9497